Charcoal activated for gas chromatography, Supelco®

Synonyms: Activated carbonCarbon activatedActivated charcoal

1.09631.0500
52428-668EA 838.24 USD
52428-668
Charcoal activated for gas chromatography, Supelco®
Charcoal activated
Formula: C
MW: 12.01 g/mol
Melting Pt: 3550 °C
Density: 2 g/cm³ (20 °C)
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
ADR: 4.2,III
REACH: 01-2119488894-16
Merck Index: 13,01818

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 3550 °C
Molar Mass 12.01 g/mol
Bulk Density 150 - 440 kg/m3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR