Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Schiff reagent for analysis of aldehydes, Supelco®

Supplier: MilliporeSigma


1.09034.1000 1.09034.1000
EM1.09034.1000CS 691.58 USD InStock
CA1.09034.1000 EM1.09034.1000
Schiff reagent for analysis of aldehydes, Supelco®
Schiff reagent
For the analytical determination of aldehydes in the ppm range.

For histology we recommend Schiff's reagent item 109033.


Order Now
Specification Test Results
Assay (SO₂) 0.15 - 0.30 %
Application test conforms


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty