Tetramethylammonium hydroxide 10% in aqueous solution Reag. Ph. Eur. for polarography, for examination of steroids, Supelco®
|
Danger
|
1.08123.0250
CA1.08123.0250
Tetramethylammonium hydroxide 10% in aqueous solution Reag. Ph. Eur. for polarography, for examination of steroids, Supelco®
Tetramethylammonium hydroxide
For polarography and for examination of steroids.
Formula:
C₄H₁₃NO MW: 91.15 g/mol Boiling Pt: 110 °C (1013 hPa) Density: 1.02 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008280 CAS Number: 75-59-2 EINECS: 200-882-9 UN: 1835 ADR: 8,II |
Specification Test Results
Assay (acidimetric) | ≥ 10.0 % |
Chloride (Cl) | ≤ 0.004 % |
Ammonia and other amines | ≤ 0.2 % |
Ag (Silver) | ≤ 0.001 % |
Fe (Iron) | ≤ 0.001 % |
Pb (Lead) | ≤ 0.001 % |
Evaporation residue | passes test |
Ash | ≤ 0.05 % |
Suitability for polarography | passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...