Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

O-Phenylenediamine for Pharma synthesis, Non-GMP

Supplier: MilliporeSigma

Danger
Synonyms: OPD1,2-Phenylendiamine1,2-Diaminobenzene1,2-Phenylenediamine

8.45071.0250 8.45071.1000 8.45071.1000 8.45071.0250
EM8.45071.0250CS 1452.96 USD InStock
EM8.45071.0250 CA8.45071.1000 EM8.45071.1000 CA8.45071.0250
O-Phenylenediamine for Pharma synthesis, Non-GMP
o-Phenylenediamine


Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 257 °C (1013 hPa)
Melting Pt: 103 °C
Density: 1.21 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007721
CAS Number: 95-54-5
EINECS: 202-430-6
UN: 1673
ADR: 6.1,III
Merck Index: 12,07438
Order Now
Specification Test Results
Solubility 54 g/l (20 °C)
Melting Point 99 - 102 °C
Molar Mass 108.14 g/mol
Bulk Density 530 kg/m3
Boiling Point 257 °C (1013 hPa)
Vapor Pressure 0.013 hPa (20 °C)
Flash Point 110 °C
Explosion Limit 1.5 %(V)
Density 1.14 g/cm3 (20 °C)
pH 7 - 8 (50 g/l, H2O, 20 °C)
Ignition Point 540 °C
Hazard Class 6.1
Hazard Class Description Toxic materials


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty