Hydroxynaphthol blue indicator for metal titration

Supplier: MilliporeSigma

Warning

Synonyms: 1-(2-Hydroxy-4-sulfo-1-naphthylazo)-2-naphthol-3,6-disulfonic acid trisodium salt1-(2-Hydroxy-4-sulpho-1-naphthylazo)-2-naphthol-3,6-disulphonic acid trisodium salt

1.04593.0025 1.04593.0025
EM1.04593.0025CS 28958.56 USD InStock
CA1.04593.0025 EM1.04593.0025
Hydroxynaphthol blue indicator for metal titration
Hydroxynaphthol blue
Formula: C₂₀H₁₁N₂Na₃O₁₁S₃
MW: 620,47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036393
CAS Number: 63451-35-4
EINECS: 264-197-7

Order Now

Specification Test Results

Appearance of solution conforms
Loss on Drying (105 °C) ≤ 5 %
Suitability as indicator (for metal titration) conforms

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR