Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Avantor is taking action. Our response to the Novel Coronavirus outbreak.

n-Hexane, dried, Seccosolv®

Supplier: MilliporeSigma

Danger
Synonyms: HexanenHexaneHexyl hydride

1.04373.0500 1.04373.0500
EM1.04373.0500CS 192.58 USD InStock
CA1.04373.0500 EM1.04373.0500
n-Hexane, dried, Seccosolv®
n-Hexane


Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69 °C (1013 hPa)
Melting Pt: –94,3 °C
Density: 0.659 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://us.vwr.com/stibo/web/std.lang.all/70/18/737018.jpg
Order Now
Specification Test Results
Solubility 0.0095 g/l (20 °C)
Melting Point -94.3 °C
Molar Mass 86.18 g/mol
Boiling Point 69 °C (1013 hPa)
Vapor Pressure 160 hPa (20 °C)
Flash Point -22 °C
Refractive Index 1.375
Explosion Limit 1.0 - 8.1 %(V)
Density 0.66 g/cm3 (20 °C)
pH (H2O) not applicable
Ignition Point 240 °C
Viscosity kinematic 0.50 mm2/s (20 °C)
Hazard Class 3
Hazard Class Description Flammable liquid


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty