Dicyclohexylcarbodiimide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: DCCN,N'-Dicyclohexylcarbodiimide1,3-Dicyclohexylcarbodiimide

M-Clarity™ quality level = MQ200

8.02954.2500 8.02954.2500 8.02954.1000 8.02954.0250 8.02954.1000 8.02954.0250
EM8.02954.2500CS 3009 USD
CA8.02954.0250 EM8.02954.2500 EM8.02954.1000 CA8.02954.1000 EM8.02954.0250 CA8.02954.2500
Dicyclohexylcarbodiimide for synthesis, Sigma-Aldrich®
Dicyclohexylcarbodiimide
Formula: C₁₃H₂₂N₂
MW: 206.33 g/mol
Boiling Pt: 148…152 °C (15 hPa)
Melting Pt: 35…36 °C
Density: 0.95 g/cm³ (40 °C)
Flash Pt: 113 °C
MDL Number: MFCD00011659
CAS Number: 538-75-0
EINECS: 208-704-1
UN: 2811
ADR: 6.1,III
Merck Index: 13,03123

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 35 - 36 °C
Molar Mass 206.33 g/mol
Bulk Density 920 kg/m3
Boiling Point 148 - 152 °C (15 hPa)
Vapor Pressure 0.0006 hPa
Flash Point 113 °C
Density 0.95 g/cm3 (40 °C)
Hazard Class 6.1
Hazard Class Description Toxic materials

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR