2-Thiobarbituric acid reagent for sorbic acid, Supelco®
Supplier: MilliporeSigma
Synonyms:
4,6-Dihydroxy-2-mercaptopyrimidine
1.08180.0025
EM1.08180.0025CS
15526
USD
EM1.08180.0025
2-Thiobarbituric acid reagent for sorbic acid, Supelco®
2-Thiobarbituric acid
Formula:
C₄H₄N₂O₂S MW: 144.15 g/mol Boiling Pt: 419 °C (1013 hPa) Melting Pt: 235 °C Density: 1.661 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006674 CAS Number: 504-17-6 EINECS: 207-985-8 |
Specification Test Results
Solubility | (20 °C) slightly soluble |
Melting Point | 235 °C |
Molar Mass | 144.14 g/mol |
Bulk Density | 210 kg/m3 |
pH | 1.7 - 1.9 (10 g/l, H2O, 20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...