Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

Eschka's mixture, GR analytical reagent, for the determination of sulphur in coal

Supplier: MilliporeSigma


1.03162.0250 1.03162.0250
EM1.03162.0250CS 3993.27 USD InStock
EM1.03162.0250 CA1.03162.0250
Eschka's mixture, GR analytical reagent, for the determination of sulphur in coal
Eschka's mixture


CAS Number: 8007-09-8
EINECS: 000-000-0
Order Now
Specification Test Results
Solubility 80 g/l (20 °C)
Bulk Density 300 kg/m3
pH 12 (50 g/l, H2O, 20 °C)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty