Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

N,N-Dimethylformamide, dried, Seccosolv®

Danger
Synonyms: Formic acid dimethylamideDimethylformamideDMFAFormdimethylamideDMFN-FormyldimethylamineFormyldimethylamine

1.02375.0161 1.02375.1001 1.02375.2500 1.02375.2500 1.02375.1001 1.02375.0161
EM1.02375.1001CS 1238.73 USD InStock
CA1.02375.0161 EM1.02375.1001 CA1.02375.2500 EM1.02375.2500 CA1.02375.1001 EM1.02375.0161
N,N-Dimethylformamide, dried, Seccosolv®
N,N-Dimethylformamide


Formula: HCON(CH₃)₂
MW: 73.1 g/mol
Boiling Pt: 153 °C (1013 hPa)
Melting Pt: -61 °C
Density: 0.949 g/cm³ (20 °C)
Flash Pt: 57.5 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://us.vwr.com/stibo/web/std.lang.all/25/06/2302506.jpg https://us.vwr.com/stibo/web/std.lang.all/06/12/16640612.jpg
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point -61 °C
Molar Mass 73.09 g/mol
Boiling Point 153 °C (1013 hPa)
Vapor Pressure 3.77 hPa (20 °C)
Flash Point 67 °C
Refractive Index 1.43
Explosion Limit 2.2 - 16 %(V)
Density 0.94 g/cm3 (20 °C)
pH 7 (200 g/l, H2O, 20 °C)
Ignition Point 410 °C
Hazard Class 3
Hazard Class Description Flammable liquid

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty