Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

Charcoal activated for AOX determination

Supplier: MilliporeSigma

Synonyms: Activated carbonCarbon activatedActivated charcoal

1.02216.0010 1.02216.0010
EM1.02216.0010CS 31926.74 USD InStock
CA1.02216.0010 EM1.02216.0010
Charcoal activated for AOX determination
Charcoal activated


Formula: C
MW: 12.01 g/mol
Melting Pt: 3550 °C
Density: 2 g/cm³ (20 °C)
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
UN: 1362
ADR: 4.2,III
REACH: 01-2119488894-16
Merck Index: 13,01818
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 3550 °C
Molar Mass 12.01 g/mol
Bulk Density 150 - 440 kg/m3


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty