Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

Brucine ≥99.5%, GR analytical reagent

Supplier: MilliporeSigma

Danger
Synonyms: 2,3-Dimethoxystrychnidin-10-one10,11-Dimethoxystrychnine

1.01952.0050 1.01952.0050
EM1.01952.0050CS 34184.55 USD InStock
EM1.01952.0050 CA1.01952.0050
Brucine ≥99.5%, GR analytical reagent
Brucine


Formula: C₂₃H₂₆N₂O₄
MW: 394.47 g/mol
MDL Number: MFCD00005942
CAS Number: 357-57-3
Order Now
Specification Test Results
Solubility (20 °C) slightly soluble
Melting Point 175 - 178 °C
Molar Mass 394.47 g/mol
Bulk Density 230 kg/m3
Hazard Class 6.1
Hazard Class Description Toxic materials


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty