Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Propanol, anhydrous (max. 0.005% H₂O) ≥99.9% (by GC), Seccosolv®

Supplier: MilliporeSigma

Danger
Synonyms: 2-HydroxypropaneIPAIsopropanolIsopropyl alcoholsec-PropanolPropan-2-olPropane, 2-hydroxy-sec-Propyl AlcoholIsopropyl Rubbing Alcohol

1.00994.0500 1.00994.0161 1.00994.1001 1.00994.0500 1.00994.0161 1.00994.1001
EM1.00994.0500CS 206.48 USD InStock
CA1.00994.0500 CA1.00994.0161 CA1.00994.1001 EM1.00994.0500 EM1.00994.0161 EM1.00994.1001
2-Propanol, anhydrous (max. 0.005% H₂O) ≥99.9% (by GC), Seccosolv®
2-Propanol


Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82.4 °C (1013 hPa)
Melting Pt: -89 °C
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://us.vwr.com/stibo/web/std.lang.all/19/69/731969.jpg
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point -89.5 °C
Molar Mass 60.1 g/mol
Boiling Point 82.4 °C (1013 hPa)
Vapor Pressure 43 hPa (20 °C)
Flash Point 12 °C
Refractive Index 1.378
Explosion Limit 2 - 13.4 %(V)
Density 0.786 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral
Ignition Point 425 °C
Evaporation number 11
Water absorption 1000 g/kg
Hazard Class 3
Hazard Class Description Flammable liquid


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty