Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

Diethyl ether, dried, Seccosolv®

Danger
Synonyms: Ether2,2'-OxybisethaneEthoxyethaneEthyl Ether2,2'-OxydiethaneEthyl ether

1.00929.0161 1.00929.0500 1.00929.1000 1.00929.1001 1.00929.0161 1.00929.1000 1.00929.1001 1.00929.0500
EM1.00929.0500CS 327.35 USD InStock
CA1.00929.0161 EM1.00929.0500 EM1.00929.1000 CA1.00929.1001 EM1.00929.0161 CA1.00929.1000 EM1.00929.1001 CA1.00929.0500
Diethyl ether, dried, Seccosolv®
Diethyl ether


Formula: (CH₃CH₂)₂O
MW: 74.12 g/mol
Boiling Pt: 35 °C (1013 hPa)
Melting Pt: -123 °C
Density: 0.7135 g/cm³ (20 °C)
Flash Pt: -40 °C
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS Number: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://us.vwr.com/stibo/web/std.lang.all/02/58/730258.jpg
Order Now
Specification Test Results
Solubility 69 g/l (20 °C)
Melting Point -116.3 °C
Molar Mass 74.12 g/mol
Boiling Point 34.6 °C (1013 hPa)
Vapor Pressure 587 hPa (20 °C)
Flash Point -40 °C
Explosion Limit 1.7 - 36 %(V)
Density 0.71 g/cm3
Ignition Point 180 °C
Hazard Class 3
Hazard Class Description Flammable liquid

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty