Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
Conical-Bottom Centrifuge Tubes, 5mL, Argos Technologies
Supplier: COLE-PARMER INSTRUMENT COMPANY
4395-47 4395-49 4395-48 4395-51 4395-46 4395-53 4395-50 4395-52
10027-562PK 71.97 USD InStock
10027-562 10027-564 10027-572 10027-568 10811-244 10027-574 10027-566 10027-570
Conical-Bottom Centrifuge Tubes, 5mL, Argos Technologies
Tubes Centrifuge Tubes
The Argos graduated 5mL prep tubes offer a convenient alternative to 15mL conical tubes for applications involving volumes less than 5mL
The 5mL prep tubes can withstand autoclaving to 120°C (248°F) for 15 minutes at 15psi, and are suitable for freezing to –90°C (–130°F). These tubes are certified Rnase-, Dnase-, and Pyrogen-free.
All tubes can be centrifuged in a FlexiFuge Mini Centrifuge, or in the same centrifuge rotor that spins standard 15mL tubes. Tubes are rated at speeds up to 14,000rpm. Easy to read graduations and a frosted marking area on the tube wall and cap are provided for sample identification.
For applications that require boiling or harsh chemicals, the T2075A CapSafe tube features a specially designed cap with a locking feature that securely snaps the tube shut. The CapSafe tube provides the same temperature range and certifications as the standard 5mL tubes.
These tubes are made of polypropylene.
The smaller tube size allows for only the pipet tip to be inserted into the tube, rather than the entire pipet barrel.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty