SP Bel-Art Safe-Lab™ Glass Stopper for Tapered Joints, Bel-Art Products, a part of SP
H20199-1330
70680-404EA 46.04 USD
70680-404
SP Bel-Art Safe-Lab™ Glass Stopper for Tapered Joints, Bel-Art Products, a part of SP
Interchangeable with penny head stoppers, this lightweight hollow borosilicate glass stopper has a flat hex head for easy gripping.
  • Lightweight
  • Hollow borosilicate glass stopper
  • Thread polypropylene extract nut allows safe removal of stopper
  • Autoclavable
Threaded polypropylene extracting nut above ground surface allows easy and safe removal of stopper from flask, and helps to eliminate accidents and breakage. Autoclave safe at 121°C (250°F).