Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
Detergents Reagent Powder Pillows, Hach
Supplier: Hach
100868 100868
10765-200PK 14.16 USD InStock
10765-200 CA100868
Detergents Reagent Powder Pillows, Hach
Environmental Test Kits
For determination of Anionic Surfactants (Detergents) by the Crystal Violet method
Hach method 8028. For use with colorimeters and spectrophotometers. Pack of 25 powder pillows.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty