Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

Water, Sterile Distilled

Supplier: GROWCELLS MS
Synonyms: Dihydrogen oxideIce

SDW-010L SDW-2000-6 SDW-2000
101076-140EA 109.33 USD InStock
101076-140 101076-138 101076-136
Water, Sterile Distilled
Water
Used the preparation of cell culture media and laboratory reagents. Cell culture quality and molecular biological grade.

Growcells provides the highest quality of Nuclease Free water for critical applications. Water is purified, tested and certified to be devoid of any activiy in DNAse, RNAse, and Protease assays. Available in convenient volumes and packaging to fit all applications requiring biologically inactive water.
Polycarbonate bottle packaging is re-usable and autoclavable.

Ordering information: TriForest Polycarbonate bottle packaging is re-usable and autoclavable.


Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
UN: 0000
Merck Index: 13,10098
Order Now
Specification Test Results
Storage RT
Shipping Ground
Shelf Life 3 Years (from Date of Manufacture)
Sterility No Growth
Activity None Detected

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty