Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Water Molecular biology grade nuclease-free, sterile

Supplier: GROWCELLS MS
Synonyms: Dihydrogen oxideIce

NUPW-0125 NUPW-2000 NUPW-0500-20 NUPW-0125-4 NUPW-010L NUPW-2000-6 NUPW-0500-2 NUPW-0125-48 NUPW-1000-12 NUPW-0500 NUPW-1000
101385-706EA 19.31 USD InStock
101385-706 101076-130 100720-998 100720-992 101076-134 101076-132 100720-996 100720-994 101076-128 101385-710 101385-712
Water Molecular biology grade nuclease-free, sterile
Water
Certified Nuclease Free water is ideal for all applications that can be sensitive to residual bi-products of DEPC breakdown. Ideal for transformation, transfection, oocyte injection and all other research applications. Water is sterile filtered and steam sterilized inside final packaging to ensure sterility.

Growcells provides the highest quality of Nuclease Free water for critical applications. Water is purified, tested and certified to be devoid of any activiy in DNAse, RNAse, and Protease assays. Available in convenient volumes and packaging to fit all applications requiring biologically inactive water.
Polycarbonate bottle packaging is re-usable and autoclavable.


Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
UN: 0000
Merck Index: 13,10098
Order Now
Specification Test Results
Sterile Filtered Yes (0.10 μM)
Storage RT
Shipping Ground
Shelf Life 3 Years (from Date of Manufacture)
Activity None Detected
Resistivity 18.2mΩ
Autoclaved Yes

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty