Glycerine (plant derived), certified

Supplier: GROWCELLS MS

Synonyms: GlysaninGlycerinGlyceritolPropane-1,2,3-triolOsmoglynPropanetriolGlycerolGlyrolGlycyl alcohol1,2,3-PropanetriolTrihydroxypropane

MRGE-4003-6 MRGE-4003 MRGE-4002
101385-684EA 71.3 USD
101385-684 101064-028 100915-642
Glycerine (plant derived), certified
Glycerine

Ultrapure plant derived undiluted glycerol suitable for use as a cryoprotectant or in various molecular biology and spectrophotometry applications.

Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
MDL Number: MFCD00004722
CAS Number: 56-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR