HQ440d Benchtop Meter Package with LBOD101 Optical BOD Probe and Hach WIMS™ BOD Manager Software, Hach
Supplier: Hach
HQDBODMGR
89501-098EA 8456.36 USD
89501-098
HQ440d Benchtop Meter Package with LBOD101 Optical BOD Probe and Hach WIMS™ BOD Manager Software, Hach
Electrochemical Meters Oxygen Meters
The Hach HQ440d benchtop meter is an advanced laboratory meter that takes the guesswork out of measurements.
  • Portable meter measures critical water quality parameters, without the need for multiple instruments
  • Intuitive user interface for simple operation and accurate results
  • Trust your measurements - IntelliCAL™ optical BOD smart probe stores all calibrations in the probe
  • Enhance productivity with an ultra-bright screen and large font size for easy to read results
  • Convenient kit includes everything you need to start testing
HQd meters connect with smart probes that automatically recognize the testing parameter, calibration history, and method settings to minimize errors and setup time. The advanced benchtop meter is designed to withstand years of continuous lab use. HQd meters feature a simple user interface that does not require manuals or training to operate. The breakthrough luminescent BOD probe has no membranes to replace for hassle-free maintenance and stabilizes quickly LDO technology and integrated stirrer. Hach Water Information Management (WIMS) BOD Manager software is included for trouble free collection, calculation, and recording of error-free BOD test results.
Ordering information: Meter package includes HQ440d Benchtop Meter, LBOD101 optical BOD probe, WIMS BOD Manager Software, Software installation support, 4 AA batteries, power adapter, USB cable for data transfer, quick-start guide, user manual, and documentation CD.