Value Master™ Natural Rubber Gloves, Showa
Supplier: Showa
VMUXL-10 VMS-07 VMUPS-07 VMUPL-09
80086-011CS 353.09 USD
80086-011 32892-085 32892-083 CA32892-082
Value Master™ Natural Rubber Gloves, Showa
Gloves
Value Master™ gloves are for manufacturing, food processing, janitorial, light-duty plant maintenance and clean up.
  • Gloves feature a rolled cuff with a tractor tread grip
  • Encourages run off of fluids often trapped by diamond-shaped embossed patterns
  • VM feature 100%-cotton flocking for added comfort
Value Master™ resists a wide range of chemicals, including animal fats, caustics, acids, oils, salts, alcohols and detergents.