SP Bel-Art Glove Box Iris Port Entry Cover, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
H50025-0009
47748-188EA 379.51 USD
47748-188
SP Bel-Art Glove Box Iris Port Entry Cover, Bel-Art Products, a part of SP
Covers
Iris port cover works with (54848-130) Glove Box Iris Port Entry.
  • Allows iris ports to be closed off when glove box is not in use