Would you like to log in again?
HQ40d Portable Meter Kit with LBOD101 Luminescent/Optical Dissolved Oxygen (LDO) Probe for BOD Measurement, Hach
Supplier: Hach
HQDBOD01 HQDBOD01
89501-096EA 2775.16 USD InStock
CA11024-770 89501-096
HQ40d Portable Meter Kit with LBOD101 Luminescent/Optical Dissolved Oxygen (LDO) Probe for BOD Measurement, Hach
Electrochemical Meters Oxygen Meters
The Hach HQ40d portable meter is an advanced meter that takes the guesswork out of measurements
  • Portable meter measures critical water quality parameters, without the need for multiple instruments
  • Intuitive user interface for simple operation and accurate results
  • Trust your measurements - IntelliCAL™ LBOD optical DO smart probe stores all calibrations in the probe
  • Designed for demanding conditions
  • Convenient kit includes everything you need to start testing
Barometric pressure measurement for automatic compensation of DO when using an LDO or LBOD probe.
HQd meters connect with smart probes that automatically recognize the testing parameter, calibration history, and method settings to minimize errors and setup time. The durable portable meter and optional rugged probes are designed to withstand years of rugged portable and field use. HQd meters feature a simple user interface that does not require manuals or training to operate.
Certifications: CE.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty