Lithium bis(trifluoromethylsulfonyl)imide ≥98.0% for synthesis, Sigma-Aldrich®

Danger

Synonyms: Lithium bis(trifluoromethanesulphonimide)Lithium Bis(Trifluoromethanesulfonyl)ImideLithium bis((trifluoromethyl)sulphonyl)amideLithium Bis(Trifluoromethanesulphonyl)ImideBis(trifluoromethane)sulfonimide lithium saltBis(trifluoromethane)sulphonimide lithium saltLithium bis((trifluoromethyl)sulfonyl)amideLithium bis(trifluoromethanesulfonimide)

M-Clarity™ quality level = MQ200

8.43873.0050 8.43873.0050
EM8.43873.0050CS 4647.48 USD
EM8.43873.0050 CA8.43873.0050
Lithium bis(trifluoromethylsulfonyl)imide ≥98.0% for synthesis, Sigma-Aldrich®
Lithium bis(trifluoromethylsulfonyl)imide
Formula: C₂F₆LiNO₄S₂
MW: 287.09 g/mol
Melting Pt: 233 °C
Density: 2.152 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00210017
CAS Number: 90076-65-6
EINECS: 415-300-0
UN: 2923
ADR: 8 (6.1),II

Order Now

Specification Test Results

Assay (alkalimetric after ionic exchange) ≥ 98.0 %
Identity (IR) passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR