Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,1,3,3-Tetramethyl-2-thiourea for synthesis

Supplier: MilliporeSigma

Synonyms: TetramethylthioureaTetramethyl-2-thioureaN,N,N',N'-Tetramethylthiourea

8.41854.0025 8.41854.0025
EM8.41854.0025CS 4619.82 USD InStock
EM8.41854.0025 CA8.41854.0025
1,1,3,3-Tetramethyl-2-thiourea for synthesis
1,1,3,3-Tetramethyl-2-thiourea


Formula: C₅H₁₂N₂S
MW: 132.23 g/mol
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 74 - 76 °C
MDL Number: MFCD00008324
CAS Number: 2782-91-4
EINECS: 220-488-0
Order Now
Specification Test Results
Solubility 32 g/l (20 °C)
Melting Point 75 - 78 °C
Molar Mass 132.22 g/mol
Boiling Point 245 °C (1013 hPa)
Vapor Pressure 1.9 hPa (25 °C)
Flash Point 100 °C


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty