N-Isopropylbenzylamine for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
N-Benzylisopropylamine
, N-Benzyl-N-isopropylamine
M-Clarity™ quality level = MQ200
8.41618.0100
8.41618.0100
EM8.41618.0100CS
1588
USD
CA8.41618.0100
EM8.41618.0100
N-Isopropylbenzylamine for synthesis, Sigma-Aldrich®
N-Isopropylbenzylamine
Formula:
C₆H₅CH₂N(CH₃)(CH₂CH₃) MW: 149.24 g/mol Boiling Pt: 200 °C (1013 hPa) Melting Pt: ∼ 0 °C Density: 0.89 g/cm³ (20 °C) Flash Pt: 87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008863 CAS Number: 102-97-6 EINECS: 203-067-6 ADR: 8,III |
Specification Test Results
Molar Mass | 149.24 g/mol |
Boiling Point | 199 - 200 °C (1013 hPa) |
Flash Point | 87 °C |
Density | 0.91 g/cm3 (20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...