Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,4-Dihydroxybenzoic acid for synthesis

Supplier: MilliporeSigma

Synonyms: Protocatechuic acid

8.41533.0025 8.41533.0100 8.41533.0100 8.41533.0025
EM8.41533.0025CS 10482.38 USD InStock
EM8.41533.0025 EM8.41533.0100 CA8.41533.0100 CA8.41533.0025
3,4-Dihydroxybenzoic acid for synthesis
3,4-Dihydroxybenzoic acid


Formula: C₇H₆O₄
MW: 154.12 g/mol
Boiling Pt: 125 - 135 °C (0.001 -rr)
Melting Pt: 202 - 204 °C
Density: 1.5415 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002509
CAS Number: 99-50-3
EINECS: 202-760-0
Merck Index: 12,08080
Order Now
Specification Test Results
Solubility 20 g/l (20 °C)
Melting Point 202 - 203 °C (decomposition)
Molar Mass 154.12 g/mol


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty