L-Norvaline for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
H-Nva-OH
, L(+)-Norvaline
, (S)-2-Aminopentanoic acid
, (S)-(+)-Norvaline
M-Clarity™ quality level = MQ200
8.41505.0001
8.41505.0001
EM8.41505.0001CS
40964
USD
EM8.41505.0001
CA8.41505.0001
L-Norvaline for synthesis, Sigma-Aldrich®
L-Norvaline
Formula:
C₅H₁₁NO₂ MW: 117.15 g/mol Boiling Pt: 228 °C (1013 hPa) Melting Pt: 101 °C |
MDL Number:
MFCD00064421 CAS Number: 6600-40-4 EINECS: 229-543-3 Merck Index: 12,06813 |
Specification Test Results
Solubility | 48.7 g/l (25 °C) |
Melting Point |
|
Molar Mass | 117.15 g/mol |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...