1,4-Diethylbenzene for synthesis, Sigma-Aldrich®

Danger


M-Clarity™ quality level = MQ200

8.41003.0005 8.41003.0005
EM8.41003.0005CS 9872.12 USD
CA8.41003.0005 EM8.41003.0005
1,4-Diethylbenzene for synthesis, Sigma-Aldrich®
1,4-Diethylbenzene
Formula: C₁₀H₁₄
MW: 134.22 g/mol
Boiling Pt: 183…184 °C (1013 hPa)
Melting Pt: –43 °C
Density: 0.862 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009264
CAS Number: 105-05-5
EINECS: 203-265-2
UN: 2049
ADR: 3,III

Order Now

Specification Test Results

Solubility 24.8 mg/l (20 °C)
Melting Point -43 °C
Molar Mass 134.22 g/mol
Boiling Point 182 - 184 °C
Vapor Pressure 1.45 hPa (25 °C)
Flash Point 55 °C
Explosion Limit 0.8 %(V)
Density 0.862 g/cm3 (20 °C)
Ignition Point 430 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR