Bis(4-fluorophenyl)methane for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
4,4'-Difluorodiphenylmethane
M-Clarity™ quality level = MQ200
8.40056.0010
8.40056.0010
EM8.40056.0010CS
47063
USD
CA8.40056.0010
EM8.40056.0010
Bis(4-fluorophenyl)methane for synthesis, Sigma-Aldrich®
Bis(4-fluorophenyl)methane
Formula:
C₁₃H₁₀F₂ MW: 204.22 |
MDL Number:
MFCD00000360 CAS Number: 457-68-1 EINECS: 207-274-2 |
Specification Test Results
Solubility | (20 °C) slightly soluble |
Molar Mass | 204.22 g/mol |
Boiling Point | 265 °C |
Density | 1.16 g/cm3 (20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...