Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Penta-O-acetyl-ɑ-D-glucopyranose for synthesis

Supplier: MilliporeSigma

Synonyms: 1,2,3,4,6-Penta-O-acetyl-α-D-glycopyranoseɑ-D-Glucose Pentaacetateα-D-Glucose pentaacetatePentaacetyl-ɑ-D-glucose1,2,3,4,6-Penta-O-Acetyl-A-D-Glycopyranose

8.40022.0005 8.40022.0005
EM8.40022.0005CS 23456.56 USD InStock
EM8.40022.0005 CA8.40022.0005
Penta-O-acetyl-ɑ-D-glucopyranose for synthesis
Penta-O-acetyl-alpha-D-glucopyranose


Formula: C₁₆H₂₂O₁₁
MW: 390,34 g/mol
MDL Number: MFCD00064071
CAS Number: 604-68-2
Order Now
Specification Test Results
Solubility 1 g/l (25 °C)
Melting Point 110 - 113 °C
Molar Mass 390.35 g/mol
Bulk Density 400 kg/m3
Flash Point 150 °C


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty