5-Hydroxysophthalic acid for synthesis, Sigma-Aldrich®

Warning

Synonyms: 5-Hydroxyisophthalic acid

M-Clarity™ quality level = MQ200

8.40013.0500 8.40013.0050 8.40013.0500 8.40013.0050
EM8.40013.0050CS 1590.14 USD
CA8.40013.0050 CA8.40013.0500 EM8.40013.0050 EM8.40013.0500
5-Hydroxysophthalic acid for synthesis, Sigma-Aldrich®
5-Hydroxysophthalic acid
Formula: C₈H₆O₅
MW: 182.13 g/mol
Boiling Pt: 386 °C (1013 hPa)
Melting Pt: 298…301 °C
MDL Number: MFCD00002515
CAS Number: 618-83-7
EINECS: 210-565-7

Order Now

Specification Test Results

Solubility 0.6 g/l (15 °C)
Melting Point 298 - 301 °C
Molar Mass 182.13 g/mol
pH  - 7 (H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR