Morpholine hydrochloride for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
Morpholinium chloride
M-Clarity™ quality level = MQ200
8.22101.0250
8.22101.0250
EM8.22101.0250CS
1049
USD
CA8.22101.0250
EM8.22101.0250
Morpholine hydrochloride for synthesis, Sigma-Aldrich®
Morpholine hydrochloride
Formula:
C₄H₁₀ClNO MW: 123.58 g/mol |
MDL Number:
MFCD00035287 CAS Number: 10024-89-2 EINECS: 233-029-4 |
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | 174 - 178 °C |
Molar Mass | 123.58 g/mol |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...