Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

1-Bromo-4-chlorobenzene for synthesis

Synonyms: 4-Bromochlorobenzene

8.21969.0005 8.21969.0250 8.21969.0005 8.21969.0250
EM8.21969.0005CS 21794.93 USD InStock
CA8.21969.0005 CA8.21969.0250 EM8.21969.0005 EM8.21969.0250
1-Bromo-4-chlorobenzene for synthesis
1-Bromo-4-chlorobenzene


Formula: C₆H₄BrCl
MW: 191.45 g/mol
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: 66 - 68 °C
Density: 1.576 g/cm³ (71 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000600
CAS Number: 106-39-8
EINECS: 203-392-3
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Solubility 0.045 g/l (25 °C)
Melting Point 64 - 67 °C
Molar Mass 191.45 g/mol
Bulk Density 900 kg/m3
Boiling Point 196 °C (1013 hPa)

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty