N-Benzylaniline for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
N-Phenylbenzylamine
, ((Phenylmethyl)amino)benzene
M-Clarity™ quality level = MQ200
8.20982.1000
8.20982.0250
8.20982.0250
8.20982.1000
EM8.20982.1000CS
2467.02
USD
EM8.20982.1000
EM8.20982.0250
CA8.20982.0250
CA8.20982.1000
N-Benzylaniline for synthesis, Sigma-Aldrich®
N-Benzylaniline
Formula:
C₆H₅CH₂NHC₆H₅ MW: 183.25 g/mol |
MDL Number:
MFCD00003018 CAS Number: 103-32-2 |
Specification Test Results
Solubility | 113 mg/l (25 °C) |
Melting Point | 33 - 36 °C |
Molar Mass | 183.26 g/mol |
Boiling Point | 306 - 307 °C (19 hPa) |
Flash Point | 113 °C |
Refractive Index | 1.6111 (26 °C, 589 nm) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...