N-phenylanthranilic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 2-(Phenylamino)benzoic acid

M-Clarity™ quality level = MQ200

8.20979.0100 8.20979.0100
EM8.20979.0100CS 1207 USD
CA8.20979.0100 EM8.20979.0100
N-phenylanthranilic acid for synthesis, Sigma-Aldrich®
N-phenylanthranilic acid
Formula: C₁₃H₁₁NO₂
MW: 213.24 g/mol
MDL Number: MFCD00002421
CAS Number: 91-40-7

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 182 - 185 °C
Molar Mass 213.23 g/mol
Bulk Density 200 kg/m3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR