Norbornadiene stabilized for synthesis, Sigma-Aldrich®

Danger

Synonyms: 2,5-Norbornadiene

M-Clarity™ quality level = MQ200

8.20918.0250
CA8.20918.0250
Norbornadiene stabilized for synthesis, Sigma-Aldrich®
Norbornadiene
Formula: C₇H₈
MDL Number: MFCD00082301
CAS Number: 121-46-0
UN: 2251
ADR: 3,II

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point -20 °C
Molar Mass 92.14 g/mol
Boiling Point 90 °C
Vapor Pressure 69 hPa (20 °C)
Flash Point -11 °C
Refractive Index 1.4702 (20 °C, 589 nm)
Explosion Limit 1.6 - 6.3 %(V)
Density 0.906 g/cm3 (20 °C)
Ignition Point 350 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR