meta-Tartaric acid for synthesis, Sigma-Aldrich®

Synonyms: m-Tartaric acid

M-Clarity™ quality level = MQ200

8.20771.0250 8.20771.0250
EM8.20771.0250CS 1740.31 USD
EM8.20771.0250 CA8.20771.0250
meta-Tartaric acid for synthesis, Sigma-Aldrich®
meta-Tartaric acid
Formula: (C₄H₄O₅)n
CAS Number: 56959-20-7

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 168 - 169 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR