2-Imidazolidinethione for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 2-ImidazolidinethioneEthlenethioureaEthylenethiourea2-Mercaptoimidazoline

M-Clarity™ quality level = MQ200

8.20696.0250 8.20696.1000 8.20696.1000 8.20696.0250
EM8.20696.0250CS 1106 USD
CA8.20696.0250 CA8.20696.1000 EM8.20696.0250 EM8.20696.1000
2-Imidazolidinethione for synthesis, Sigma-Aldrich®
2-Imidazolidinethione
Formula: C₃H₆N₂S
MW: 102.16 g/mol
MDL Number: MFCD00005276
CAS Number: 96-45-7

Order Now

Specification Test Results

Solubility 8 g/l (20 °C)
Melting Point 196 - 199 °C
Molar Mass 102.16 g/mol
Bulk Density 750 - 900 kg/m3
Vapor Pressure 0.2 hPa (170 °C)
Flash Point 253 °C
Density 1.4 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR