4'-Hydroxypropiophenone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Paroxypropione4’-Hydroxypropiophenonep-Hydroxypropiophenone

M-Clarity™ quality level = MQ200

8.20692.0100 8.20692.0100
EM8.20692.0100CS 1161 USD
CA8.20692.0100 EM8.20692.0100
4'-Hydroxypropiophenone for synthesis, Sigma-Aldrich®
4'-Hydroxypropiophenone
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
MDL Number: MFCD00002361
CAS Number: 70-70-2

Order Now

Specification Test Results

Solubility 0.345 g/l (15 °C)
Melting Point 148 - 151 °C
Molar Mass 150.18 g/mol
Bulk Density 300 - 400 kg/m3
Flash Point 112 °C
Density 1.09 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR