Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

Dimethyl fumarate for synthesis

Supplier: MilliporeSigma

Synonyms: Fumaric acid dimethyl ester

8.20583.0250 8.20583.0100 8.20583.0100 8.20583.0250
EM8.20583.0250CS 3349.33 USD InStock
EM8.20583.0250 EM8.20583.0100 CA8.20583.0100 CA8.20583.0250
Dimethyl fumarate for synthesis
Dimethyl fumarate


Formula: C₆H₈O₄
MW: 144,13 g/mol
Boiling Pt: 192 °C (1013 hPa)
Melting Pt: 102 °C
Density: 1.37 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00064438
CAS Number: 624-49-7
EINECS: 210-849-0
Merck Index: 13,04308
Order Now
Specification Test Results
Solubility 1.6 g/l (20 °C)
Melting Point 102 °C
Molar Mass 144.13 g/mol
Boiling Point 193 °C (1013 mbar)
Vapor Pressure 5 hPa (25 °C)
Density 1.37 g/cm3
pH 4.0 - 6.0 (10 g/l, H2O, 20 °C) suspension


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty