N,N-Diethylaniline for synthesis, Sigma-Aldrich®

Danger


M-Clarity™ quality level = MQ200

8.20395.2500 8.20395.2500 8.20395.1000 8.20395.0100 8.20395.1000 8.20395.0100
EM8.20395.1000CS 299.54 USD
CA8.20395.1000 CA8.20395.0100 CA8.20395.2500 EM8.20395.1000 EM8.20395.0100 EM8.20395.2500
N,N-Diethylaniline for synthesis, Sigma-Aldrich®
N,N-Diethylaniline
Formula: C₁₀H₁₅N
MW: 149.24 g/mol
MDL Number: MFCD00009042
CAS Number: 91-66-7
UN: 2432
ADR: 6.1,III

Order Now

Specification Test Results

Solubility 0.13 g/l (20 °C)
Melting Point -38.8 °C
Molar Mass 149.23 g/mol
Boiling Point 217 °C (1013 hPa)
Vapor Pressure 0.2 hPa (20 °C)
Flash Point 79 °C
Refractive Index 1.5409 (20 °C, 589 nm)
Explosion Limit 1.1 - 5.3 %(V)
Density 0.93 g/cm3 (20 °C)
pH 8 (0.1 g/l, H2O, 20 °C)
Ignition Point 500 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR