Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4,4'-Bis(dimethylamino)thiobenzophenone for synthesis

Supplier: MilliporeSigma


8.20162.0010 8.20162.0010
EM8.20162.0010CS 11438.85 USD InStock
CA8.20162.0010 EM8.20162.0010
4,4'-Bis(dimethylamino)thiobenzophenone for synthesis
4,4'-Bis(dimethylamino)thiobenzophenone


Formula: C₁₇H₂₀N₂S
MW: 284,43 g/mol
CAS Number: 1226-46-6
EINECS: 214-962-6
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 198 - 206 °C
Molar Mass 284.42 g/mol


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty