1-Bromotetradecane for synthesis, Sigma-Aldrich®

Synonyms: Myristyl bromide

M-Clarity™ quality level = MQ200

8.18896.0250 8.18896.0005 8.18896.0250 8.18896.0005
EM8.18896.0005CS 2911.4 USD
CA8.18896.0250 EM8.18896.0005 CA8.18896.0005 EM8.18896.0250
1-Bromotetradecane for synthesis, Sigma-Aldrich®
1-Bromotetradecane
Formula: BrH₂C(CH₂)₁₂CH₃
MW: 277.29 g/mol
MDL Number: MFCD00000228
CAS Number: 112-71-0

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 4 - 6 °C
Molar Mass 277.28 g/mol
Boiling Point 175 - 178 °C (27 hPa)
Flash Point 149 °C
Refractive Index 1.4575 (20 °C, 589 nm)
Density 1.02 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR