Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

Tetrapropylammonium bromide for synthesis

Supplier: MilliporeSigma


8.18735.0250 8.18735.0100 8.18735.0250 8.18735.0100
EM8.18735.0100CS 1536 USD InStock
CA8.18735.0250 EM8.18735.0100 EM8.18735.0250 CA8.18735.0100
Tetrapropylammonium bromide for synthesis
Tetrapropylammonium bromide


Formula: C₁₂H₂₈BrN
MW: 266,27 g/mol
Melting Pt: 265 to 274 °C
MDL Number: MFCD00011840
CAS Number: 1941-30-6
EINECS: 217-727-6
Order Now
Specification Test Results
Solubility 1000 g/l (20 °C)
Melting Point 265 - 274 °C (decomposition)
Molar Mass 266.26 g/mol
pH 5 - 7 (100 g/l, H2O)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty