4-Chloro-3,5-xylenol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Chloroxylenol4-Chloro-3,5-dimethylphenol2-Chloro-5-hydroxy-3-xylene

M-Clarity™ quality level = MQ200

8.18734.0250
EM8.18734.0250CS 425 USD
EM8.18734.0250
4-Chloro-3,5-xylenol for synthesis, Sigma-Aldrich®
4-Chloro-3,5-xylenol
Formula: C₈H₉ClO
MW: 156.61 g/mol
MDL Number: MFCD00002324
CAS Number: 88-04-0

Order Now

Specification Test Results

Solubility 250 mg/l (20 °C)
Melting Point 114 - 116 °C
Molar Mass 156.61 g/mol
Boiling Point 246 °C (1013 hPa)
Flash Point 132 °C
Density 0.67 g/cm3
pH 7 (0.2 g/l, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR