(R)-(+)-Pulegone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: p-Menth-4(8)-en-3-one(R)-p-Menth-4(8)-en-3-one(R)-5-Methyl-2-(propan-2-ylidene)cyclohexanone(R)-2-Isopropylidene-5-methylcyclohexanone(+)-Pulegone

M-Clarity™ quality level = MQ200

8.18665.0010 8.18665.0010
EM8.18665.0010CS 181045 USD
EM8.18665.0010 CA8.18665.0010
(R)-(+)-Pulegone for synthesis, Sigma-Aldrich®
(R)-(+)-Pulegone
Formula: C₁₀H₁₆O
MW: 152.23 g/mol
MDL Number: MFCD00063000
CAS Number: 89-82-7

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Molar Mass 152.23 g/mol
Boiling Point 97 °C (15.99 mbar)
Vapor Pressure 0.16 hPa (25 °C)
Flash Point 82 °C
Refractive Index 1.4864 (20 °C, 589 nm)
Density 0.935 g/m3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR