2-(tert-Butylamino)ethanol for synthesis, Sigma-Aldrich®

Danger


M-Clarity™ quality level = MQ200

8.18661.0101
CA8.18661.0101
2-(tert-Butylamino)ethanol for synthesis, Sigma-Aldrich®
2-(tert-Butylamino)ethanol
Formula: C₆H₁₅NO
MW: 117.19 g/mol
MDL Number: MFCD00002837
CAS Number: 4620-70-6
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 40 - 43 °C
Molar Mass 117.19 g/mol
Boiling Point 176 - 177 °C (1013 hPa)
Flash Point 69 °C
Density 0.86 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR