Glycidyl isopropyl ether for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 2,3-Epoxypropyl Isopropyl Ether

M-Clarity™ quality level = MQ200

8.18598.2500 8.18598.2500 8.18598.0100
EM8.18598.2500EA 404 USD
CA8.18598.2500 CA8.18598.0100 EM8.18598.2500
Glycidyl isopropyl ether for synthesis, Sigma-Aldrich®
Glycidyl isopropyl ether
Formula: C₆H₁₂O₂
UN: 1992
ADR: 3 (6.1),III

Order Now

Specification Test Results

Solubility 188 g/l (19 °C)
Molar Mass 116.15 g/mol
Boiling Point 137 °C (1013 hPa)
Vapor Pressure 13 hPa (25 °C)
Flash Point 33 °C
Refractive Index 1.4101 (20 °C, 589 nm)
Density 0.92 g/cm3 (20 °C)
pH 7 (188 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR